1-(5-Nitro-2-pyridinyl)-4-piperidinecarboxylic acid

Lieferant: Apollo Scientific

OR8480-1G OR8480-5G
APOSOR8480-1GEA 154 EUR
APOSOR8480-1G APOSOR8480-5G
1-(5-Nitro-2-pyridinyl)-4-piperidinecarboxylic acid
1-(5-Nitro-2-pyridinyl)-4-piperidinecarboxylic acid
Formel: C₁₁H₁₃N₃O₄
Molecular Weight: 251.24 g/mol
Schmelzpunkt: 182…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06200945
CAS-Nummer: 868077-44-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR