Benzyl-2-isopropyl-4-oxotetrahydro-1(2H)-pyridinecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Isopropylpiperidin-4-one, N-CBZ protected
OR8488-1G
OR8488-250MG
APOSOR8488-1GEA
350
EUR
APOSOR8488-1G
APOSOR8488-250MG
Benzyl-2-isopropyl-4-oxotetrahydro-1(2H)-pyridinecarboxylate
Benzyl-2-isopropyl-4-oxotetrahydro-1(2H)-pyridinecarboxylate
Formel:
C₁₆H₂₁NO₃ Molecular Weight: 275.35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437612 CAS-Nummer: 952183-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...