tert-Butyl-3-(4-chlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate
Lieferant: Apollo Scientific
Synonyme:
3-(4-Chlorobenzyl)-3-formylpiperidine, N-BOC protected
OR8495-1G
OR8495-500MG
APOSOR8495-1GEA
489
EUR
APOSOR8495-1G
APOSOR8495-500MG
tert-Butyl-3-(4-chlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate
tert-Butyl-3-(4-chlorobenzyl)-3-formyltetrahydro-1(2H)-pyridinecarboxylate
Formel:
C₁₈H₂₄ClNO₃ Molecular Weight: 337,85 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09607940 CAS-Nummer: 952183-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...