Ethyl-2-(1-methyl-2,5-dioxotetrahydro-4H-imidazol-4-yliden)acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl (2,5-dioxo-1-methylimidazolidin-4-ylidene)acetate

OR8509-1G OR8509-5G
APOSOR8509-1GEA 198 EUR
APOSOR8509-1G APOSOR8509-5G
Ethyl-2-(1-methyl-2,5-dioxotetrahydro-4H-imidazol-4-yliden)acetate
Ethyl-2-(1-methyl-2,5-dioxotetrahydro-4H-imidazol-4-yliden)acetate
Formel: C₈H₁₀N₂O₄
Molecular Weight: 198,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09607892
CAS-Nummer: 712-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR