3-(4-Ethylphenyl)-3-oxopropanenitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR8510-1G
OR8510-250MG
OR8510-5G
APOSOR8510-1GEA
183
EUR
APOSOR8510-1G
APOSOR8510-250MG
APOSOR8510-5G
3-(4-Ethylphenyl)-3-oxopropanenitrile
3-(4-Ethylphenyl)-3-oxopropanenitrile
Formel:
C₁₁H₁₁NO Schmelzpunkt: 105…107 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02260779 CAS-Nummer: 96220-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...