Methyl-5-(4-bromophenoxy)-2-furoate

Lieferant: Apollo Scientific

OR8515-1G OR8515-5G
APOSOR8515-1GEA 270 EUR
APOSOR8515-1G APOSOR8515-5G
Methyl-5-(4-bromophenoxy)-2-furoate
Methyl-5-(4-bromophenoxy)-2-furoate
Formel: C₁₂H₉BrO₄
Molecular Weight: 297,11 g/mol
Schmelzpunkt: 64…66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607928
CAS-Nummer: 952183-41-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR