5-[(4-Methylpiperazino)methyl]-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-5-[(4-methylpiperazin-1-yl)methyl]-2-thioxopyrimidin-4(1H)-one

OR8518-10G OR8518-1G OR8518-5G
APOSOR8518-10GEA 546 EUR
APOSOR8518-10G APOSOR8518-1G APOSOR8518-5G
5-[(4-Methylpiperazino)methyl]-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone
5-[(4-Methylpiperazino)methyl]-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone
Formel: C₁₀H₁₆N₄OS
Molecular Weight: 240,33 g/mol
Schmelzpunkt: 227…228 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09607894
CAS-Nummer: 952183-05-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR