5-Amino-2,4-dichlorophenol

Lieferant: Apollo Scientific
Gefahr

OR8523-10G OR8523-1G OR8523-5G
APOSOR8523-10GEA 677 EUR
APOSOR8523-10G APOSOR8523-1G APOSOR8523-5G
5-Amino-2,4-dichlorophenol
5-Amino-2,4-dichlorophenol
Formel: C₆H₅Cl₂NO
Molecular Weight: 178,012 g/mol
Schmelzpunkt: 133…137 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00186187
CAS-Nummer: 39489-79-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR