5-Brom-4-methoxy-2-(methylthio)pyrimidin
Lieferant: Apollo Scientific
OR8528-5G
OR8528-1G
APOSOR8528-5GEA
429
EUR
APOSOR8528-5G
APOSOR8528-1G
5-Brom-4-methoxy-2-(methylthio)pyrimidin
5-Bromo-4-methoxy-2-(methylthio)pyrimidine
Formel:
C₆H₇BrN₂OS Lagertemperatur: Raumtemperatur |
CAS-Nummer:
81560-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...