6-Brom-1,3-dichlorisochinolin 96%

Lieferant: Apollo Scientific
Gefahr

OR8568-1G OR8568-5G OR8568-10G
APOSOR8568-1GEA 77 EUR
APOSOR8568-1G APOSOR8568-5G APOSOR8568-10G
6-Brom-1,3-dichlorisochinolin 96%
6-Bromo-1,3-dichloroisoquinoline
Formel: C₉H₄BrCl₂N
Molecular Weight: 276,9448 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09258639
CAS-Nummer: 552331-05-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR