6-Chlorindol

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloro-1H-indole

OR8577-5G OR8577-100G OR8577-25G
APOSOR8577-5GEA 21.9 EUR
APOSOR8577-5G APOSOR8577-100G APOSOR8577-25G
6-Chlorindol
6-Chlorindol
Formel: C₈H₆ClN
Molecular Weight: 151.6 g/mol
Schmelzpunkt: 87…91 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005681
CAS-Nummer: 17422-33-2
EINECS: 241-449-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR