3-Chlor-4-hydroxyphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Chloro-4-hydroxyphenyl)boronic acid

OR8583-1G OR8583-5G
APOSOR8583-1GEA 21.9 EUR
APOSOR8583-1G APOSOR8583-5G
3-Chlor-4-hydroxyphenylboronsäure 98%
3-Chlor-4-hydroxyphenylboronsäure
Formel: C₆H₆BClO₃
Molecular Weight: 172,3700 g/mol
Schmelzpunkt: 223…228 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09258739
CAS-Nummer: 182344-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR