4-Brom-1H-pyrazol 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR8592-100G
OR8592-25G
OR8592-500G
OR8592-250G
APOSOR8592-100GEA
24.8
EUR
APOSOR8592-100G
APOSOR8592-25G
APOSOR8592-500G
APOSOR8592-250G
4-Brom-1H-pyrazol 98%
4-Bromo-1H-pyrazole
Formel:
C₃H₃BrN₂ Molecular Weight: 146,97 g/mol Siedepunkt: 250…260 °C Schmelzpunkt: 93…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075602 CAS-Nummer: 2075-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...