2-(Boc-amino)-5-brompyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Boc-2-Amino-5-bromopyridine, 2-Amino-5-bromopyridine, 2-BOC protected

OR8593-25G OR8593-5G OR8593-100G
APOSOR8593-25GEA 49.3 EUR
APOSOR8593-25G APOSOR8593-5G APOSOR8593-100G
2-(Boc-amino)-5-brompyridin 98%
2-(Boc-amino)-5-brompyridin
Formel: C₁₀H₁₃BrN₂O₂
Molecular Weight: 273,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06411381
CAS-Nummer: 159451-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR