4-Amino-3-chlor-5-methylbenzonitril 98%

Lieferant: Apollo Scientific
Achtung

OR8613-5G OR8613-25G
APOSOR8613-5GEA 109 EUR
APOSOR8613-5G APOSOR8613-25G
4-Amino-3-chlor-5-methylbenzonitril 98%
4-Amino-3-chlor-5-methylbenzonitril
Formel: C₈H₇ClN₂
Molecular Weight: 166.61 g/mol
Schmelzpunkt: 128…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041476
CAS-Nummer: 158296-69-6
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR