6-Brom-2-naphthylamin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromonaphthalen-2-amine, 2-Amino-6-bromonaphthalene
OR8620-1G
OR8620-25G
OR8620-5G
APOSOR8620-1GEA
105
EUR
APOSOR8620-1G
APOSOR8620-25G
APOSOR8620-5G
6-Brom-2-naphthylamin
6-Brom-2-naphthylamin
Formel:
C₁₀H₈BrN Molecular Weight: 222.0812 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01026466 CAS-Nummer: 7499-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...