(Phenylthio)propanone 97%

Lieferant: Apollo Scientific
Achtung

OR8634-25G OR8634-5G
APOSOR8634-25GEA 314 EUR
APOSOR8634-25G APOSOR8634-5G
(Phenylthio)propanone 97%
(Phenylthio)propanone
Formel: C₉H₁₀OS
Molecular Weight: 166,24 g/mol
Schmelzpunkt: 40…43 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00014992
CAS-Nummer: 5042-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR