2-Piperidinoisonicotinonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR8649-1G
OR8649-250MG
APOSOR8649-1GEA
53.8
EUR
APOSOR8649-1G
APOSOR8649-250MG
2-Piperidinoisonicotinonitril 97%
2-Piperidinoisonicotinonitril
Formel:
C₁₁H₁₃N₃ Schmelzpunkt: 55…57,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200992 CAS-Nummer: 127680-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...