Methyl-5-bromonicotinoylacetate 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 3-(5-bromopyridin-3-yl)-3-oxopropanoate
OR8693-1G
OR8693-5G
APOSOR8693-1GEA
139
EUR
APOSOR8693-1G
APOSOR8693-5G
Methyl-5-bromonicotinoylacetate 95+%
Methyl-5-bromonicotinoylacetate
Formel:
C₉H₈BrNO₃ Molecular Weight: 258,08 g/mol Schmelzpunkt: 93…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216527 CAS-Nummer: 205985-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...