7-Chloro-4-(piperazin-1-yl)quinoline 97%

Lieferant: Apollo Scientific
Gefahr

OR8697-1G OR8697-250MG
APOSOR8697-1GEA 29 EUR
APOSOR8697-1G APOSOR8697-250MG
7-Chloro-4-(piperazin-1-yl)quinoline 97%
7-Chloro-4-(piperazin-1-yl)quinoline
Formel: C₁₃H₁₄ClN₃
Molecular Weight: 247,7234 g/mol
Schmelzpunkt: 113…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01476123
CAS-Nummer: 837-52-5
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR