3-[Benzyl(tert-butoxycarbonyl)amino]propanoic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Benzylamino)propanoic acid, N-BOC protected
OR8718-1G
OR8718-5G
APOSOR8718-1GEA
261
EUR
APOSOR8718-1G
APOSOR8718-5G
3-[Benzyl(tert-butoxycarbonyl)amino]propanoic acid 95+%
3-[Benzyl(tert-butoxycarbonyl)amino]propanoic acid
Formel:
C₁₅H₂₁NO₄ Molecular Weight: 279,34 g/mol Schmelzpunkt: 48…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037900 CAS-Nummer: 289889-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...