(R)-(+)-4-Isopropyl-2-oxazolidinon

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-4-Isopropyloxazolidin-2-one, (4R)-(+)-4-Isopropyl-2-oxazolidinone

OR8729-25G OR8729-5G
APOSOR8729-25GEA 104 EUR
APOSOR8729-25G APOSOR8729-5G
(R)-(+)-4-Isopropyl-2-oxazolidinon
(R)-(+)-4-Isopropyl-2-oxazolidinon
Formel: C₆H₁₁NO₂
Molecular Weight: 129.16 g/mol
Schmelzpunkt: 70…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075172
CAS-Nummer: 95530-58-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR