(S)-(-)-4-Benzyl-2-oxazolidinon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-4-Benzyl-2-oxazolidinone, (S)-4-Benzyloxazolidin-2-one
OR8747-100G
OR8747-25G
OR8747-500G
APOSOR8747-100GEA
55.1
EUR
APOSOR8747-100G
APOSOR8747-25G
APOSOR8747-500G
(S)-(-)-4-Benzyl-2-oxazolidinon
(S)-(-)-4-Benzyl-2-oxazolidinon
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177,2 g/mol Schmelzpunkt: 86…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064496 CAS-Nummer: 90719-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...