4-(2-Keto-1-benzimidazolinyl)piperidine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1,3-Dihydro-1-(piperidin-4-yl)-2H-benzimidazol-2-one
OR8751-1G
OR8751-5G
OR8751-10G
APOSOR8751-1GEA
81.2
EUR
APOSOR8751-1G
APOSOR8751-5G
APOSOR8751-10G
4-(2-Keto-1-benzimidazolinyl)piperidine 97%
4-(2-Keto-1-benzimidazolinyl)piperidine
Formel:
C₁₂H₁₅N₃O Molecular Weight: 217,272 g/mol Schmelzpunkt: 183…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005714 CAS-Nummer: 20662-53-7 EINECS: 243-950-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...