4-(4-Bromphenyl)-4-piperidinol 97%

Lieferant: Apollo Scientific

Synonyme: 4-(4'-Bromophenyl)-4-hydroxypiperidine

OR8753-1G OR8753-5G
APOSOR8753-1GEA 82.9 EUR
APOSOR8753-1G APOSOR8753-5G
4-(4-Bromphenyl)-4-piperidinol 97%
4-(4-Bromphenyl)-4-piperidinol
Formel: C₁₁H₁₄BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00067034
CAS-Nummer: 57988-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR