4-Chloro-2,6-dimethyl-3-nitropyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR8763-1G
OR8763-5G
OR8763-10G
APOSOR8763-1GEA
21.9
EUR
APOSOR8763-1G
APOSOR8763-5G
APOSOR8763-10G
4-Chloro-2,6-dimethyl-3-nitropyridine
4-Chloro-2,6-dimethyl-3-nitropyridine
Formel:
C₇H₇ClN₂O₂ Schmelzpunkt: 70…73 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08543468 CAS-Nummer: 15513-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...