2-Nitrobenzolsulfonamid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Nitrobenzolsulfonsäureamid
OR8788-100G
OR8788-25G
OR8788-5G
APOSOR8788-100GEA
87
EUR
APOSOR8788-100G
APOSOR8788-25G
APOSOR8788-5G
2-Nitrobenzolsulfonamid 98%
2-Nitrobenzolsulfonamid
Formel:
C₆H₆N₂O₄S Molecular Weight: 202,19 g/mol Schmelzpunkt: 188…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009807 CAS-Nummer: 5455-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...