D(+)-α-Acetobromglucose

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide, Bromo-2,3,4,6-Tetra-o-Acetyl-α-D-Glucopyranose, Acetobromo glucose, (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, α-Acetobromoglucose, Acetobromo-α-D-glucose

OR8800T-25G OR8800T-5G
APOSOR8800T-25GEA 156 EUR
APOSOR8800T-25G APOSOR8800T-5G
D(+)-α-Acetobromglucose
D(+)-α-Acetobromglucose
Formel: C₁₄H₁₉BrO₉
Molecular Weight: 411,2 g/mol
Schmelzpunkt: 89…90 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00063254
CAS-Nummer: 572-09-8
EINECS: 209-339-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR