4-Hydroxy-3,5-diiodbenzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Diiodo4-hydroxybenzaldehyde, 3,5-Diiodo-4-hydroxybenzaldehyde
OR8809-25G
OR8809-5G
APOSOR8809-25GEA
63.8
EUR
APOSOR8809-25G
APOSOR8809-5G
4-Hydroxy-3,5-diiodbenzaldehyd
4-Hydroxy-3,5-diiodbenzaldehyd
Formel:
C₇H₄I₂O₂ Molecular Weight: 373,92 g/mol Schmelzpunkt: 202…203 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014670 CAS-Nummer: 1948-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...