3,5-Diiodsalicylaldehyd 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Diiodo-2-hydroxybenzaldehyde

OR8810-100G
APOSOR8810-100GEA 127 EUR
APOSOR8810-100G
3,5-Diiodsalicylaldehyd 98%
3,5-Diiodsalicylaldehyd
Formel: C₇H₄I₂O₂
Molecular Weight: 373,92 g/mol
Schmelzpunkt: 105…107 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00003321
CAS-Nummer: 2631-77-8
EINECS: 220-117-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR