2-Mercaptoimidazol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Imidazol-2-thiol, 2-Mercapto-1H-imidazol
OR8823-1G
OR8823-5G
OR8823-10G
APOSOR8823-1GEA
26.1
EUR
APOSOR8823-1G
APOSOR8823-5G
APOSOR8823-10G
2-Mercaptoimidazol 97%
2-Mercaptoimidazole
Formel:
C₃H₄N₂S Molecular Weight: 100,14 g/mol Schmelzpunkt: 228…231 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005188 CAS-Nummer: 872-35-5 EINECS: 212-823-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...