3,4-Dichlorpyridin

Lieferant: Apollo Scientific
Achtung

OR8832-5G OR8832-10G
APOSOR8832-5GEA 21.9 EUR
APOSOR8832-5G APOSOR8832-10G
3,4-Dichlorpyridin
3,4-Dichloropyridine
Formel: C₅H₃Cl₂N
Molecular Weight: 147.99 g/mol
Schmelzpunkt: 22…23 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01861989
CAS-Nummer: 55934-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR