Methyl-phenoxyacetat 98%
Lieferant: Apollo Scientific
Synonyme:
Phenoxyacetic Acid Methyl Ester, Methyl 2-phenoxyacetate
OR8842-500G
APOSOR8842-500GEA
127
EUR
APOSOR8842-500G
Methyl-phenoxyacetat 98%
Methyl-phenoxyacetat
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010227 CAS-Nummer: 2065-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...