Methylmalonylchlorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Methyl 3-chloro-3-oxopropanoate
OR8845-100G
OR8845-25G
OR8845-250G
APOSOR8845-100GEA
119
EUR
APOSOR8845-100G
APOSOR8845-25G
APOSOR8845-250G
Methylmalonylchlorid 97%
Methylmalonylchlorid
Formel:
C₄H₅ClO₃ Molecular Weight: 136,53 g/mol Siedepunkt: 57…59 °C/12mmHg Dichte: 1,273 g/cm³ (25 °C) Flammpunkt: 80 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00013657 CAS-Nummer: 37517-81-0 EINECS: 253-540-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...