Coumarin-6-carboxaldehyde 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR8856-1G
OR8856-5G
APOSOR8856-1GEA
72.5
EUR
APOSOR8856-1G
APOSOR8856-5G
Coumarin-6-carboxaldehyde 95+%
Coumarin-6-carboxaldehyde
Formel:
C₁₀H₆O₃ Schmelzpunkt: 192…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01941322 CAS-Nummer: 51690-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...