2-Chlor-3-thiophenboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Chlorothiophen-3-yl)boronic acid

OR8869-1G OR8869-5G
APOSOR8869-1GEA 62.5 EUR
APOSOR8869-1G APOSOR8869-5G
2-Chlor-3-thiophenboronsäure
2-Chloro-3-thiopheneboronic acid
Formel: C₄H₄BClO₂S
Molecular Weight: 162,4 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD05662391
CAS-Nummer: 177734-82-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR