3-Brom-4-chloranisol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-chloro-4-methoxybenzene

OR8887-25G OR8887-5G OR8887-100G
APOSOR8887-25GEA 88.7 EUR
APOSOR8887-25G APOSOR8887-5G APOSOR8887-100G
3-Brom-4-chloranisol
3-Brom-4-chloranisol
Formel: C₇H₆BrClO
Molecular Weight: 221,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070739
CAS-Nummer: 2732-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR