Norlaudanosine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride, Tetrahydropapaverine Hydrochloride
OR8892-1G
OR8892-25G
OR8892-5G
APOSOR8892-1GEA
21.9
EUR
APOSOR8892-1G
APOSOR8892-25G
APOSOR8892-5G
Norlaudanosine hydrochloride
Norlaudanosine hydrochloride
Formel:
C₂₀H₂₅NO₄ Molecular Weight: 379,88 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035267 CAS-Nummer: 6429-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...