5-Brom-2-methoxyphenol
Lieferant: Apollo Scientific
|
Achtung
|
OR8907-100G
OR8907-25G
APOSOR8907-100GEA
156
EUR
APOSOR8907-100G
APOSOR8907-25G
5-Brom-2-methoxyphenol
5-Brom-2-methoxyphenol
Formel:
C₇H₇BrO₂ Molecular Weight: 203,0300 g/mol Siedepunkt: 150 °C/20mmHg Schmelzpunkt: 63…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00673137 CAS-Nummer: 37942-01-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...