8-Chinolincarbonsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: Quinoline-8-carboxylic acid

OR8958-1G OR8958-5G OR8958-10G
APOSOR8958-1GEA 26.1 EUR
APOSOR8958-1G APOSOR8958-5G APOSOR8958-10G
8-Chinolincarbonsäure
8-Chinolincarbonsäure
Formel: C₁₀H₇NO₂
Molecular Weight: 173,17 g/mol
Schmelzpunkt: 183…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047619
CAS-Nummer: 86-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR