N-Boc-ethylamine 98%

Lieferant: Apollo Scientific

Synonyme: tert-Butyl N-ethylcarbamate, tert-Butyl ethylcarbamate

OR900318-5G
APOSOR900318-5GEA 151 EUR
APOSOR900318-5G
N-Boc-ethylamine 98%
N-Boc-ethylamine
Formel: C₇H₁₅NO₂
Molecular Weight: 145,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12923020
CAS-Nummer: 38267-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR