N-Methyl-p-toluolsulfonamid 98%

Lieferant: Apollo Scientific

Synonyme: N,4-Dimethylbenzenesulphonamide

OR900480-100G OR900480-500G
APOSOR900480-100GEA 33.5 EUR
APOSOR900480-100G APOSOR900480-500G
N-Methyl-p-toluolsulfonamid 98%
N-Methyl-p-toluolsulfonamid
Formel: C₈H₁₁NO₂S
Molecular Weight: 185,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008285
CAS-Nummer: 640-61-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR