Boc-N-Me-Dl-Val-Oh 97%
Lieferant: Apollo Scientific
Synonyme: Boc-N-methyl-DL-valine
OR900527-1G OR900527-5G
APOSOR900527-1GEA 126 EUR
APOSOR900527-1G APOSOR900527-5G
Boc-N-Me-Dl-Val-Oh 97%
Boc-N-Me-Dl-Val-Oh
Formel: C₁₁H₂₁NO₄
Molecular Weight: 231.292 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00153317
CAS-Nummer: 13850-91-4