Propoxybenzene 98%

Lieferant: Apollo Scientific

Synonyme: Phenyl propyl ether

OR900706-1G OR900706-5G
APOSOR900706-1GEA 33.5 EUR
APOSOR900706-1G APOSOR900706-5G
Propoxybenzene 98%
Propoxybenzene
Formel: C₉H₁₂O
Molecular Weight: 136,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039935
CAS-Nummer: 622-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR