Ethyl isoxazole-4-carboxylate 95%

Lieferant: Apollo Scientific

OR900850-1G OR900850-250MG
APOSOR900850-1GEA 174 EUR
APOSOR900850-1G APOSOR900850-250MG
Ethyl isoxazole-4-carboxylate 95%
Ethyl isoxazole-4-carboxylate
Formel: C₆H₇NO₃
Molecular Weight: 141.13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368244
CAS-Nummer: 80370-40-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR