2-(1-Pyrrolidinyl)ethanol 98%

Lieferant: Apollo Scientific

Synonyme: 1-Pyrrolidineethanol, N-(2-Hydroxyethyl)pyrrolidine

OR901254-100G OR901254-500G
APOSOR901254-100GEA 78.3 EUR
APOSOR901254-100G APOSOR901254-500G
2-(1-Pyrrolidinyl)ethanol 98%
1-(2-Hydroxyethyl)pyrrolidine
Formel: C₆H₁₃NO
Molecular Weight: 115,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003181
CAS-Nummer: 2955-88-6
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR