2-Butyrylpyridine 98%
Lieferant: Apollo Scientific
Synonyme:
1-(Pyridin-2-yl)butan-1-one
OR901609-1G
OR901609-25G
OR901609-5G
APOSOR901609-1GEA
88.6
EUR
APOSOR901609-1G
APOSOR901609-25G
APOSOR901609-5G
2-Butyrylpyridine 98%
2-Butyrylpyridine
Formel:
C₉H₁₁NO Molecular Weight: 149.19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00778319 CAS-Nummer: 22971-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...