2-Cyano-3-methoxypyridine 98%

Lieferant: Apollo Scientific

Synonyme: 3-Methoxypicolinonitrile, 3-methoxypyridine-2-carbonitrile

OR901628-1G OR901628-5G
APOSOR901628-1GEA 145 EUR
APOSOR901628-1G APOSOR901628-5G
2-Cyano-3-methoxypyridine 98%
2-Cyano-3-methoxypyridine
Formel: C₇H₆N₂O
Molecular Weight: 134,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07374875
CAS-Nummer: 24059-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR