2,8-Dichlorchinolin 98%
Lieferant: Apollo Scientific
OR901662-25G
APOSOR901662-25GEA
142
EUR
APOSOR901662-25G
2,8-Dichlorchinolin 98%
2,8-Dichloroquinoline
Formel:
C₉H₅Cl₂N Molecular Weight: 198,0500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00832077 CAS-Nummer: 4470-83-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...