2-Chlor-6,8-dimethylchinolin 98%

Lieferant: Apollo Scientific

OR901674-1G OR901674-25G OR901674-5G
APOSOR901674-1GEA 125 EUR
APOSOR901674-1G APOSOR901674-25G APOSOR901674-5G
2-Chlor-6,8-dimethylchinolin 98%
2-Chloro-6,8-dimethylquinoline
Formel: C₁₁H₁₀ClN
Lagertemperatur: Raumtemperatur
MDL: MFCD15143624
CAS-Nummer: 139719-23-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR